Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589345.2 | XP_005589402.1 | 3 - 4 | 1128 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |