Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005556371.2 | XP_005556428.1 | 1 | 273 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |