Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STPG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575947.2 | XP_005576004.2 | 4 - 5 | 939 | 62 | |
XM_015433691.1 | XP_015289177.1 | 3 - 4 | 1742 | 62 | ||
XM_015433692.1 | XP_015289178.1 | 3 - 4 | 1744 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |