Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015448891.1 | XP_015304377.1 | 1 | 1513 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |