Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285203.1 | NP_001272132.1 | 10 - 11 | 1226 | 77 | |
GenBank mRNA | AB168877.1 | - | 10 - 11 | 1226 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |