Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590617.2 | XP_005590674.1 | 2 - 3 | 2376 | 100 | |
XM_005590618.2 | XP_005590675.1 | 2 - 3 | 1424 | 100 | ||
XM_005590621.2 | XP_005590678.1 | 2 - 3 | 1368 | 100 | ||
XM_005590622.2 | XP_005590679.1 | 2 - 3 | 1521 | 100 | ||
XM_015441299.1 | XP_015296785.1 | 2 - 3 | 1638 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |