Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283622.1 | NP_001270551.1 | 1 | 1121 | 96 | |
GenBank mRNA | AB169045.1 | - | 1 | 1121 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |