Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283923.1 | NP_001270852.1 | 4 - 5 | 1056 | 126 | |
XM_015440960.1 | XP_015296446.1 | 5 - 6 | 853 | 126 | ||
XM_015440962.1 | XP_015296448.1 | 5 - 6 | 879 | 126 | ||
XM_015440963.1 | XP_015296449.1 | 4 - 5 | 1070 | 126 | ||
XM_015440964.1 | XP_015296450.1 | 5 - 6 | 876 | 126 | ||
GenBank mRNA | AB060218.1 | - | 4 - 5 | 1056 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |