Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284640.1 | NP_001271569.1 | 678 | 97 | ||
GenBank mRNA | AB097521.1 | - | 678 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most |