Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285271.1 | NP_001272200.1 | 2 - 3 | 405 | 84 | |
XM_005576506.2 | XP_005576563.1 | 3 - 4 | 611 | 84 | ||
XM_015433453.1 | XP_015288939.1 | 2 - 3 | 1122 | 84 | ||
GenBank mRNA | AB169586.1 | - | 2 - 3 | 405 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |