Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287717.1 | NP_001274646.1 | 1 - 2 | 193 | 98 | |
GenBank mRNA | AB189711.1 | - | 1 - 2 | 193 | 98 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Probe spans exons |