Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283409.1 | NP_001270338.1 | 11 - 12 | 1159 | 68 | |
XM_005568801.2 | XP_005568858.1 | 11 - 12 | 1094 | 68 | ||
XM_015457630.1 | XP_015313116.1 | 11 - 12 | 2085 | 68 | ||
GenBank mRNA | AB083314.2 | - | 11 - 12 | 1159 | 68 | |
AB171304.1 | - | 11 - 12 | 1159 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |