Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285354.1 | NP_001272283.1 | 2 - 3 | 490 | 76 | |
XM_005557474.2 | XP_005557531.1 | 2 - 3 | 464 | 76 | ||
XM_005557475.2 | XP_005557532.1 | 2 - 3 | 387 | 76 | ||
XM_005557476.2 | XP_005557533.1 | 2 - 3 | 233 | 76 | ||
GenBank mRNA | AB168797.1 | - | 2 - 3 | 490 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |