Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283676.1 | NP_001270605.1 | 7 - 8 | 968 | 102 | |
XM_005547965.2 | XP_005548022.1 | 7 - 8 | 889 | 102 | ||
XM_005547967.2 | XP_005548024.1 | 7 - 8 | 934 | 102 | ||
XM_015445279.1 | XP_015300765.1 | 7 - 8 | 863 | 102 | ||
GenBank mRNA | AB168592.1 | - | 7 - 8 | 971 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |