Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEKT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284892.1 | NP_001271821.1 | 3 - 4 | 385 | 72 | |
XM_005543977.2 | XP_005544034.1 | 3 - 4 | 712 | 72 | ||
XM_005543978.2 | XP_005544035.1 | 3 - 4 | 357 | 72 | ||
GenBank mRNA | AB169442.1 | - | 3 - 4 | 387 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |