Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285145.1 | NP_001272074.1 | 4 - 5 | 378 | 86 | |
GenBank mRNA | AB179473.1 | - | 4 - 5 | 379 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |