Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285190.1 | NP_001272119.1 | 2 - 3 | 132 | 174 | |
XM_015437796.1 | XP_015293282.1 | 2 - 3 | 234 | 174 | ||
GenBank mRNA | AB168504.1 | - | 2 - 3 | 132 | 174 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |