Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285317.1 | NP_001272246.1 | 13 - 14 | 2307 | 101 | |
XM_005544665.2 | XP_005544722.1 | 13 - 14 | 2376 | 101 | ||
XM_005544667.1 | XP_005544724.1 | 12 - 13 | 2268 | 101 | ||
GenBank mRNA | AB071053.1 | - | 1664 | 101 | ||
AB168993.1 | - | 13 - 14 | 2307 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |