Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284126.1 | NP_001271055.1 | 3 - 4 | 463 | 81 | |
GenBank mRNA | AB169566.1 | - | 3 - 4 | 465 | 81 | |
AB170084.1 | - | 3 - 4 | 463 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |