Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283096.1 | NP_001270025.1 | 4 - 5 | 780 | 104 | |
XM_005550638.2 | XP_005550695.1 | 5 - 6 | 1309 | 104 | ||
XM_005550640.2 | XP_005550697.1 | 4 - 5 | 1278 | 104 | ||
XM_015447303.1 | XP_015302789.1 | 4 - 5 | 715 | 104 | ||
GenBank mRNA | AB169093.1 | - | 4 - 5 | 780 | 104 | |
AB169129.1 | - | 2 - 3 | 177 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |