Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283222.1 | NP_001270151.1 | 4 - 5 | 857 | 83 | |
XM_015442156.1 | XP_015297642.1 | 3 - 4 | 1614 | 83 | ||
GenBank mRNA | AB169655.1 | - | 4 - 5 | 858 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |