Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283353.1 | NP_001270282.1 | 5 - 6 | 897 | 70 | |
XM_005552323.2 | XP_005552380.1 | 6 - 7 | 1126 | 70 | ||
GenBank mRNA | AB168917.1 | - | 6 - 7 | 1063 | 70 | |
AB179453.1 | - | 5 - 6 | 897 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |