Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284989.1 | NP_001271918.1 | 10 - 11 | 1345 | 93 | |
XM_005584713.2 | XP_005584770.1 | 9 - 10 | 1272 | 93 | ||
XM_015437842.1 | XP_015293328.1 | 10 - 11 | 1425 | 93 | ||
XM_015437843.1 | XP_015293329.1 | 8 - 9 | 1105 | 93 | ||
GenBank mRNA | AB169157.1 | - | 10 - 11 | 1345 | 93 | |
AB169223.1 | - | 8 - 9 | 1097 | 93 | ||
AB170341.1 | - | 9 - 10 | 1263 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |