Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284989.1 | NP_001271918.1 | 8 - 9 | 1063 | 98 | |
XM_005584713.2 | XP_005584770.1 | 7 - 8 | 990 | 98 | ||
XM_015437842.1 | XP_015293328.1 | 8 - 9 | 1143 | 98 | ||
XM_015437843.1 | XP_015293329.1 | 6 - 7 | 823 | 98 | ||
GenBank mRNA | AB169157.1 | - | 8 - 9 | 1063 | 98 | |
AB169223.1 | - | 6 - 7 | 815 | 98 | ||
AB170341.1 | - | 7 - 8 | 981 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |