Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285245.1 | NP_001272174.1 | 4 - 5 | 898 | 107 | |
XM_005560075.2 | XP_005560132.1 | 4 - 5 | 1380 | 107 | ||
XM_005560076.2 | XP_005560133.1 | 5 - 6 | 1118 | 107 | ||
XM_015452452.1 | XP_015307938.1 | 5 - 6 | 1134 | 107 | ||
XM_015452453.1 | XP_015307939.1 | 5 - 6 | 1059 | 107 | ||
GenBank mRNA | AB168338.1 | - | 4 - 5 | 898 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |