Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283870.1 | NP_001270799.1 | 1 - 2 | 211 | 75 | |
XM_015455894.1 | XP_015311380.1 | 3 - 4 | 584 | 75 | ||
GenBank mRNA | AB220471.1 | - | 1 - 2 | 211 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |