Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284066.1 | NP_001270995.1 | 6 - 7 | 693 | 69 | |
XM_005583485.2 | XP_005583542.1 | 6 - 7 | 730 | 69 | ||
GenBank mRNA | AB168985.1 | - | 6 - 7 | 693 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |