Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284649.1 | NP_001271578.1 | 5 - 6 | 891 | 80 | |
XM_005544776.2 | XP_005544833.1 | 5 - 6 | 1612 | 80 | ||
XM_005544778.2 | XP_005544835.1 | 5 - 6 | 959 | 80 | ||
GenBank mRNA | AB125182.1 | - | 5 - 6 | 891 | 80 | |
AB174536.1 | - | 5 - 6 | 891 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |