Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283290.1 | NP_001270219.1 | 2 - 3 | 361 | 84 | |
XM_015455918.1 | XP_015311404.1 | 2 - 3 | 183 | 84 | ||
GenBank mRNA | DQ074805.1 | - | 2 - 3 | 361 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |