Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015449302.1 | XP_015304788.1 | 24 - 25 | 5627 | 108 | |
XM_015449307.1 | XP_015304793.1 | 24 - 25 | 5627 | 108 | ||
XM_015449309.1 | XP_015304795.1 | 23 - 24 | 5485 | 108 | ||
XM_015449313.1 | XP_015304799.1 | 24 - 25 | 4894 | 108 | ||
XM_015449318.1 | XP_015304804.1 | 17 - 18 | 4682 | 108 | ||
XM_015449320.1 | XP_015304806.1 | 24 - 25 | 4895 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |