Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005546276.2 | XP_005546333.1 | 3 - 4 | 564 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |