Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAL3ST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005574907.2 | XP_005574964.1 | 2 - 3 | 165 | 97 | |
XM_005574908.2 | XP_005574965.1 | 2 - 3 | 604 | 97 | ||
XM_015433127.1 | XP_015288613.1 | 2 - 3 | 1432 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |