Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579485.2 | XP_005579542.1 | 2 - 3 | 355 | 76 | |
XM_005579486.2 | XP_005579543.1 | 2 - 3 | 355 | 76 | ||
XM_015435652.1 | XP_015291138.1 | 2 - 3 | 338 | 76 | ||
XM_015435653.1 | XP_015291139.1 | 2 - 3 | 338 | 76 | ||
XM_015435654.1 | XP_015291140.1 | 2 - 3 | 338 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |