Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589595.2 | XP_005589652.1 | 1 - 2 | 1456 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |