Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFIKKN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590774.2 | XP_005590831.1 | 1 - 2 | 1989 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |