Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGIX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015443323.1 | XP_015298809.1 | 2 - 3 | 367 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |