Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543322.1 | XP_005543379.1 | 4 - 5 | 1924 | 61 | |
XM_015435641.1 | XP_015291127.1 | 3 - 4 | 1761 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |