Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005548301.2 | XP_005548358.1 | 3 - 4 | 852 | 83 | |
XM_005548303.2 | XP_005548360.1 | 3 - 4 | 1380 | 83 | ||
GenBank mRNA | AB168577.1 | - | 3 - 4 | 1311 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |