Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566299.2 | XP_005566356.1 | 4 - 5 | 1339 | 98 | |
XM_015456615.1 | XP_015312101.1 | 4 - 5 | 904 | 98 | ||
XM_015456616.1 | XP_015312102.1 | 4 - 5 | 1378 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |