Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015434649.1 | XP_015290135.1 | 6 - 7 | 2871 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |