Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583676.2 | XP_005583733.2 | 1 - 2 | 907 | 72 | |
XM_015438249.1 | XP_015293735.1 | 3 - 4 | 1132 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |