Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015451705.1 | XP_015307191.1 | 3 - 4 | 299 | 67 | |
XM_015451706.1 | XP_015307192.1 | 5 - 6 | 1595 | 67 | ||
XM_015451707.1 | XP_015307193.1 | 5 - 6 | 613 | 67 | ||
XM_015451708.1 | XP_015307194.1 | 4 - 5 | 1472 | 67 | ||
XM_015451709.1 | XP_015307195.1 | 3 - 4 | 1273 | 67 | ||
XM_015451710.1 | XP_015307196.1 | 4 - 5 | 490 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |