Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102145189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015452018.1 | XP_015307504.1 | 1 | 7643 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |