Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592783.2 | XP_005592840.1 | 3 - 4 | 418 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |