Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005546620.2 | XP_005546677.1 | 4 - 5 | 2237 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |