Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005564363.2 | XP_005564420.1 | 1 - 2 | 536 | 107 | |
XM_015455391.1 | XP_015310877.1 | 2 - 3 | 578 | 107 | ||
XM_015455392.1 | XP_015310878.1 | 2 - 3 | 506 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |