Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005580502.2 | XP_005580559.1 | 4 - 5 | 721 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |