Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015437101.1 | XP_015292587.1 | 1 - 2 | 5389 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |