Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592885.2 | XP_005592942.1 | 3 - 4 | 391 | 83 | |
XM_005592886.2 | XP_005592943.1 | 3 - 4 | 464 | 83 | ||
XM_005592887.2 | XP_005592944.1 | 3 - 4 | 383 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |