Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592885.2 | XP_005592942.1 | 4 - 5 | 573 | 75 | |
XM_005592886.2 | XP_005592943.1 | 4 - 5 | 646 | 75 | ||
XM_005592887.2 | XP_005592944.1 | 4 - 5 | 565 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |